KIMBLE® NMR Sample Tubes, 5 mm, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE®
897193-7100 897193-0008 897241-0008 897130-0007 897110-0007 897110-0008 897130-0008 897225-0000 897205-0009 897245-3008 897240-0009 897200-0008 897120-0008 897150-0008 897240-0000 897225-0009 897235-0009 897200-0000 897250-3000 897240-0008 897193-8100 897245-3000 897140-0008 897200-0009 897230-0000 897120-0007 897140-0007 897235-0000 897193-0000 897220-0000 897220-0008 897235-0008 897241-0009 897193-0050 897205-0008 897241-0000 897225-0008 897150-0007 897205-0000 897230-0008 897193-8050
KT897140-0008PK 1156.05 MXN
KT897130-0007 KT897205-0008 KT897241-0000 KT897205-0009 KT897130-0008 KT897245-3000 KT897230-0008 KT897150-0008 KT897150-0007 KT897205-0000 KT897250-3000 KT897245-3008 KT897193-7100 KT897225-0000 KT897110-0008 KT897110-0007 KT897225-0009 KT897193-8050 KT897241-0009 KT897225-0008 KT897241-0008 KT897193-0050 KT897230-0000 KT897193-0008 KT897220-0000 KT897200-0000 KT897140-0008 KT897140-0007 KT897240-0008 KT897240-0009 KT897200-0009 KT897200-0008 KT897193-8100 KT897235-0008 KT897235-0009 KT897240-0000 KT897193-0000 KT897235-0000 KT897120-0007 KT897120-0008 KT897220-0008
KIMBLE® NMR Sample Tubes, 5 mm, DWK Life Sciences
Tubos Tubos NMR
Designed for high resolution work with NMR spectrometers.
  • Disposable-grade tubes are suitable for applications where sample retrieval is not required
  • High-quality tubes are ideal for sealing directly to vacuum manifolds, joints, or valves
  • Tubes feature a sandblasted marking spot
Tubes are manufactured from ASTM Type 1 borosilicate glass under rigid quality control to meet exacting specifications for wall chambers and variation. T.I.R. is the Total Indicator Reading. Each NMR tube is supplied with a polyethylene cap. Disposable and thrift-grade tubes do not have caps attached; high-quality tubes are supplied with caps attached.
Información para pedidos: Tubes for 800 and 900 MHz NMR are also available; contact your Avantor sales representative for more information.