Spectra/Por® Dialysis Tubing, Regenerated Cellulose, MWCO 12000 to 14000, Spectrum® Laboratories
Proveedor: Spectrum
Spectra/Por®
S732706 S732703 S732697 S732700 S732709
25225-248CS 96579.99 MXN
25225-260 25225-281 25225-248 25225-204 25225-226
Spectra/Por® Dialysis Tubing, Regenerated Cellulose, MWCO 12000 to 14000, Spectrum® Laboratories
Tuberías Tuberías para diálisis
Standard dialysis tubing for all laboratory dialysis applications.
The tubing contains glycerin, water, and small amounts of sulfur compounds (approx. 0.1%).