Electrode and Probe Refill Solutions, Beckman Coulter®
Proveedor: Beckman Coulter
A58726 A51713 A60217 598943 A58729 A60217 511469 A58726
BKA60217EA 4219.68 MXN
CABKA60217 BK511469 BKA58726 BKA58729 CABK598943 CABKA51713 CABKA58726 BKA60217
Electrode and Probe Refill Solutions, Beckman Coulter®
Electrolitos Electrolitos de relleno
These solutions are for use with electrodes and probes.
  • Compatible with Beckman Coulter Electrodes
  • For General Purpose, Calomel, and Low-Ionic Electrodes
  • Works with Dissolved Oxygen Probes
Solutions are compatible with Beckman Coulter® electrodes and probes.
Solutions for FUTURA™ electrodes are not compatible and should not be used with the new line of electrodes whose catalog numbers begin with BKA.