Infectious Substance Shipper, UN Marked, Qorpak®
Proveedor: Qorpak
HMS-69010
16455-260CS 19236.89 MXN
16455-260
Infectious Substance Shipper, UN Marked, Qorpak®
Remitentes de envíos y contenedores
Certified packaging for shipment of infectious substances.
Certified for Class 6.2 Infectious Substances. Acceptable for ground and air shipment.
Complete with corrugated outer carton, diecut foam, absorbent material, metal sealing ring, plastic bag and tape. Will accept up to 12 vials (vials not included).