Conductivity Cells, YSI 3400 Series
Proveedor: YSI
603440 603402 099821 603403 099820 603401
23201-078EA 26460.96 MXN
23201-045 23201-012 23201-078 23201-125 23201-103 23201-205
Conductivity Cells, YSI 3400 Series
Electrodos Sensores de conductividad
For use with YSI conductive instruments as well as with instruments of other manufacturers.
These precision cells contain platinized platinum-iridium electrodes. 23201-125 has nickel-rod electrodes. Conductivity cells require cell adapter 66121-968 (sold separately) when used with YSI Models 3100 and 3200 conductivity meters. Choose cells made from Pyrex® 7740 or ABS plastic.
Note: Cell constant values are calibrated within 1% of cell constant.