VWR® Sample Mailing Tubes
Proveedor: VWR
VWR®
3811-800-008
16465-262CS 4159.4 MXN
16465-262
VWR® Sample Mailing Tubes
Remitentes de envíos y contenedores
These economical, disposable tubes feature graduations in 1 mL increments and are suitable for shipping.
  • Leakproof
  • Frosted writing surface allows convenient labeling and identification
  • Tubes are autoclavable
Top × Bottom O.D. 15.9 x 14.6 mm (5/8 x 9/16")
Caps are made of high-density polyethylene. Tubes are made of durable polypropylene with a conical bottom and skirted base.
Tubes will withstand use in temperatures as low as –80°C. They may not be used with liquid nitrogen or nitrogen vapor.