ThermoSafe® Durable Insulated Containers, Sonoco ThermoSafe
Proveedor: Sonoco
ThermoSafe®
870 861 859 860 857 858
89496-032EA 21100.46 MXN
10818-630 89496-032 10818-628 10818-636 10818-634 10818-632
ThermoSafe® Durable Insulated Containers, Sonoco ThermoSafe
Refrigerantes Arcón hielo seco
ThermoSafe Durable Insulated Containers maintain critical temperatures for your laboratory specific materials and dry ice storage.
  • Maintain critical temperatures
  • Popular design
  • Compact to maneuver in tight spaces
  • Strong and durable
These rugged temperature sensitive bulk storage containers are the most popular design for the transport and storage of dry ice and temperature-sensitive specimens
Ranging in size and style, these durable transport chests, also known as fish boxes, make storing, transporting, and unloading efficient and worry-free. High impact polyethylene can handle the rigors of transport, and the tight fitting lid and container construction maintains temperature and maximizes transporting efficiency.
ThermoSafe insulated chests and containers are strong, durable, and specifically manufactured to be used and reused. Replacement latches, hinges, handles, and other handling and replacement accessories are available at a low cost. Contact your Avantor Account Manager for more information.
They are uniquely compact and designed to maneuver in tight spaces. Double-wall polyethylene construction filled with urethane foam insulation maintains dry ice quality.