VWR® Signature™ Aerosol Filter Pipette Tips
Proveedor: VWR
VWR® Signature™
1055-965-316 1057-965-316 1054-800-000 1059-800-000 1055-800-010 1057-800-010 1097-965-306 1059-965-306
53510-102CS 13824.21 MXN
53510-014 53510-102 89003-420 53510-016 16466-004 53510-106 16466-008 53510-094
VWR® Signature™ Aerosol Filter Pipette Tips
Puntas de pipetas
VWR® Signature™ aerosol filter pipette tips are natural barrier aerosol tips that incorporate a special high recovery polyethylene filter that traps aerosols and prevents cross-contamination by keeping the pipettor barrel clean.
  • For use with most pipettors
  • Available in sterile and non sterile formats
Sterile tips are gamma-irradiated to guarantee product sterility. Autoclavable at 15 psi, 122 °C (252 °F) for 15 minutes.
Certificaciones: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
Envase: Multiple packaging options.