Air Monitor Cassettes, 37 mm, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva
4336 4339-N 4338 4338 4336 4339-N
28145-393PK 6487.13 MXN
28145-393 CA28145-393 28145-430 28145-397 CA28145-397 CA28145-430
Air Monitor Cassettes, 37 mm, Cytiva (Formerly Pall Lab)
Filtros Filtros de casetes
Ideal for general particulate air sampling when fitted with the appropriate membrane or filter.
  • Designed with NIOSH requirements in mind
  • Ideal for air sampling
  • Styrene acrylonitrile housing
Styrene acrylonitrile housing consists of inlet, outlet, and inner ring for open- or closed-face monitoring. Cassette 10277-992 is a solvent-resistant version made with polypropylene housing.
Certificaciones: Designed to meet NIOSH requirements for industrial hygiene sampling.
Información para pedidos: Assembled unit 28145-393 includes 0.8 µm GN-4 Metricel® (mixed cellulose esters) membrane with cellulose support pad. Unassembled units do not include support pad or membrane. The 2-piece unit 28145-397 includes the styrene acrylonitrate housing only.