VWR® General Purpose, Oil-only, and Hazmat Pads and Rolls
Proveedor: VWR
VWR®
MAT900VWR MAT701VWR VWR430 MAT800VWR VWR818 MAT801VWR MAT901VWR MAT700VWR
89169-002CS 2393.62 MXN
89169-002 82028-022 82028-020 82028-026 82028-024 82028-018 89169-000 82028-028
VWR® General Purpose, Oil-only, and Hazmat Pads and Rolls
Materiales para contención de derrames
Absorbent pads and rolls are ideal for spill cleanup; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety.
  • Soak up leaks and spills fast, keeping floors at your workplace clean, dry, and safe
  • High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
  • Can be incinerated after use to reduce waste
  • Absorbents soaks up oil, water-based liquids, petroleum products, and solvents
  • Hazmat absorbents soak up aggressive fluids such as acids, caustics, and bases
General-purpose pads and rolls absorb water-based liquids, petroleum products, and solvents for everyday maintenance and cleanup. Oil-only pads and rolls absorb oils but repel water. Hazmat pads and rolls are designed for aggressive fluids such as acids, caustics, and bases.