Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene® MF75™
290-3345 291-3320 292-3320 292-4520 291-4545 291-4520 291-3345 290-3320 290-4545 290-4520
28199-307CS 2604.1 MXN
28199-296 28199-298 28199-307 28199-309 28199-303 28199-317 28199-305 28199-300 28199-302 28199-312
Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Filtros Filtros de la parte superior de botellas
SFCA (surfactant-free cellulose acetate) membrane filters are much cleaner than standard cellulose acetate and are designed for vacuum filtration with glass media bottles.
  • Certified radiation-sterilized, non-pyrogenic, and non-cytotoxic
  • Quick-disconnect sidearm accepts ¹/₄ to ³/₈" tubing
  • Use the 0.2 µm size for sterilization of aqueous solutions and culture media
Polystyrene bottle-top design features molded graduations and a support plate that minimizes foaming and eliminates the need for a separate filter pad. Filter unit screws directly onto media bottles. Yellow collar filter cup is labeled with lot number, catalog number, pore size, and expiration date.
Select the 0.45 µm size for particle analysis, scintillation counting, prefiltration of aqueous solutions, and general cytology.
Precaución: Use only with sterile bottles designed for vacuum use. Always use safety shield during vacuum procedures. For laboratory use only, not for in-vitro diagnosis or parenterals.