Nalgene® Rapid-Flow™ Tissue Culture Filter Units, Nylon Membrane, Sterile, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene® MF77™
163-0020 164-0020 154-0045 153-0045 151-4045 150-0020 150-0045 153-0020 154-0020 151-4020
28199-428CS 6405.7 MXN
28199-428 28199-417 28199-382 28199-360 28199-371 28199-359 28198-514 28199-338 28199-349 28198-510
Nalgene® Rapid-Flow™ Tissue Culture Filter Units, Nylon Membrane, Sterile, Thermo Scientific
Filtros Filtros de la parte superior de botellas
Units are designed for applications requiring low extractables.
  • Graduated polystyrene upper chamber and receiver storage bottle
  • Side arm features a cellulosic vent plug and quick-disconnect tubing adapter
  • Radiation sterile, Certified non-pyrogenic and non-cytotoxic
Naturally hydrophilic, tissue-culture grade nylon membrane exhibits less than 0.5% extractables by weight, contains no wetting agents, and is alcohol-resistant. Filter cup is printed with catalog number, lot number, membrane material, pore size, and expiration date.
They feature 1¹/₂ - turn threaded screw closure that guarantees a leakproof seal and eliminates pH shift in receiver.
Información para pedidos: Shipped complete with filter, tubing adapter, vent plug, receiver, and dust cover. Units with 75mm dia. membrane include 12 glass prefilters.
Precaución: For laboratory use only. Not for in vitro diagnosis or parenterals.