VWR® 96-Well Real Time PCR Plates
Proveedor: VWR, part of Avantor
3963-521-300
83009-674
VWR® 96-Well Real Time PCR Plates
Opaque black or white polypropylene PCR plates and optically clear cap closures are suitable for Real Time PCR applications.
  • Plates are compatible with most thermal cyclers and are ideal for high-throughput screening thermocycler applications
  • Smooth, thin, uniform well walls ensure accurate thermal transfer
  • Wells are slightly raised to accept optically clear strip caps or sealing films
  • All plates are autoclavable and certified free of RNase, DNase, and human DNA (lot-by-lot certificates available)
Thin, flexible, easy-to-cut plates have a working capacity of 200µL per well. The half-skirted plate is easy to insert and extract. Skirted plates provide reduced plate distortion during high-throughput screening and sit flat for greater stability.