Microtiter® 96-Well Plates, Thermo Scientific
Proveedor: Thermo Scientific
Microtiter®
2401 2605 2801 9205 2305-11 2205 2101
62402-907CS 4723.84 MXN
62402-918 62402-907 62402-908 62402-914 62402-910 62402-933 62402-912
Microtiter® 96-Well Plates, Thermo Scientific
Microplacas Microplacas para inmunología
Developed for general applications, including sample storage, sample or reagent transfer, serial dilution, reagent mixing, hemagglutination, and complement fixation.
These 96-well plates are available in rigid styrene or flexible vinyl. The vinyl plates may be cut into strips or individual wells, and are recommended for incubation procedures up to 60°C (140°F). Plate 62402-907 has an alphanumeric grid to aid in sample identification.