Immulon® Microtiter™ 96-Well Plates and Strips, Thermo Scientific
Proveedor: Thermo Scientific
Immulon® Microtiter™
6309 3655 6404 3355 3855 6310 6405 6302 6301 3555 3455
62402-961CS 8008.72 MXN
62403-074 62403-082 62402-947 62402-959 62403-076 62402-954 62402-966 62402-961 62402-972 62402-963 62402-952
Immulon® Microtiter™ 96-Well Plates and Strips, Thermo Scientific
Microplacas Microplacas para inmunología
These plates are ideal for a variety of colorimetric techniques, such as ELISA, which are performed in a 96-well format
Immulon 1 B exhibits a consistent medium binding affinity to proteins and complexes primarily with hydrophobic tendencies. Immulon 2 HB is an irradiated plate that provides higher binding affinity. Immulon 4 HBX features a unique resin that offers maximum protein uptake with enhanced precision and guaranteed coefficient of variation (CV) of <5%.
Choose from plates, Removawell™ Assemblies and Strips for custom formatting, and Dividastrips™ Assemblies with 2 x 8 strips that separate into two 1x8 strips.
Immulon polystyrene plastics provide a consistent surface for reproducible results over a variety of binding affinities.
Plate dimensions: 12.8 x 8.6 cm (5 x 33/8")
Información para pedidos: For Removawell Holder that holds eight 12-well or twelve 8-well Removawell strips and Removawell Workstation, see Microtiter® Accessories for 96-Well Plates, Thermo Scientific series.