Finnpipette™ Novus Multichannel Pipettors, Variable Volume, Thermo Scientific
Proveedor: Thermo Scientific
Finnpipette™
46300200 46300500 46300400 46300700 46300100 46300000 46300300 46300800
89082-250EA 52291.68 MXN
89082-252 89082-250 89082-256 89082-244 89082-254 89082-242 89082-248 89082-246
Finnpipette™ Novus Multichannel Pipettors, Variable Volume, Thermo Scientific
Pipetas Micropipetas
Pipettors combine simplicity, comfort, and functionality in an exceptionally intuitive instrument to improve productivity, precision, and ergonomic operation in liquid handling applications.
  • Pipettors feature 10 pipetting functions and nine pipetting speeds
  • Lightweight, long life battery
Pipettors are designed for easy in-lab calibration and servicing, and have a fully autoclavable tip cone to ensure sterility and avoid cross-contamination. The two-level intuitive menu and fully graphical text display provide simple control. A multilingual graphical interface is offered in seven languages: English, French, German, Italian, Japanese, Spanish, and Swedish. Unique backlight technology distinguishes the display by eliminating surrounding light reflections and improving contrast. The result is a graphical display with excellent clarity, even in low light conditions such as safety cabinets and fume hoods. The battery can be fully recharged in only one hour, while a long, unobtrusive cable permits use of the instrument during recharging. Additionally, the pipettor can be personalized with its own name, which is displayed on the screen when the instrument is not in use.
The lightweight construction helps to prevent upper body stress and strain, allowing the user to work longer without fatigue. Features include a solid handle shaped to fit comfortably and securely in the hand and a finger rest that can adjust over 120° for optimal positioning.