Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Proveedor: Thermo Scientific
Nunc® MicroWell™
442587 266120 249570 268152 260844 260836 237108 249570 260895 262146 262146 269620 268152 260836 260844 268152 269620 260895 266120 442587 237108
62409-114CS 13254.1 MXN
CA62409-108 73520-518 CA73520-506 62409-108 73520-506 CA62409-114 62409-114 CA62409-068 62407-180 62409-068 CA62407-182 CA62407-180 62407-182 62407-166 82031-368 CA82031-368 CA62407-166 CATX73520-506 CA73520-486 73520-486 CA43300-451
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Microplacas Microplacas HTS
These nonsterile 96-well plates are ideal for assays, colorimetric serology, and bacteriology applications
All plates are made of high-quality polystyrene to ensure optimum optical clarity, with the exception of plate 62409-108, which is made of polypropylene. Plate 260844 is generic bar coded.
Información para pedidos: Plate 62407-166 is provided with a lid. For lids and sealing tapes, see the 62409-118 series.