Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Proveedor: Thermo Scientific
Nunc® MicroWell™
260860 268200 262162 269787 243656 269787 260860 243656 249662 262162 268200
62407-174CS 28134.68 MXN
CA62407-174 CA62407-184 62407-174 CA62409-116 62409-116 CA62409-111 62409-112 62409-111 CA62409-120 62409-120 62407-184
Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Microplacas Microplacas HTS
These sterile, polystyrene plates are ideal for bacteriology, serology, molecular biology, and other laboratory applications
Plates have a standard format, which makes them compatible with plate washers and readers. V-bottom and U-bottom plates allow pelleting or settling of cells for gentle concentration and sample retrieval. Flat-bottom wells can be easily examined microscopically.
The wells have raised rims to reduce risk of cross-contamination. Lids have rings to control condensation during incubation. The beveled corners on the plates and lids prevent accidental reversal of the lid onto the plate. Wells are alphanumerically-labeled for easy identification. They stack conveniently and securely, with or without lids. Clone selection and replication can be performed using these plates with a few simple movements using a transferable solid phase (TSP) replicator.
Used for dilutions and general storage.
Información para pedidos: For lids and sealing tapes, see the Nunc® Microplate Lids, Thermo Scientific series.