Nunc® MicroWell™ 96-Well Plates, Polypropylene, High Volume, Thermo Scientific
Proveedor: Thermo Scientific
Nunc® MicroWell™
249950 249950 267407 267350 249943 249947 249945 249946 267245 249945 249949 267342 267334 249947 267342 249949 249946 267334 249944 249944 267245 249943 267350 267385 267407 267385
62408-936CS 20080.24 MXN
CA46600-670 CA46600-672 46600-664 CA46600-674 46600-666 CA46600-676 46600-668 CA62408-940 62408-936 CA62408-944 62408-938 CA62408-946 CA62408-948 CA46600-668 46600-672 46600-674 46600-676 CA46600-664 CA46600-666 62408-940 62408-944 46600-670 62408-946 CA62408-936 62408-948 CA62408-938
Nunc® MicroWell™ 96-Well Plates, Polypropylene, High Volume, Thermo Scientific
Microplacas Microplacas HTS
Polypropylene plates have a standard 96-well format and are designed to fit standard automation equipment and robotics.
Black plates reduce background fluorescence and minimize back-scattered light.
White plates offer maximum signal reflection and minimize autoluminescence and autofluorescence signals.
Natural plates are ideal for screening, liquid phase assays and storage. They are also useful for combinatorial chemistry applications.
Blue, red, yellow, and natural plates exhibit minimum binding for hydrophilic molecules. They are ideal for sample collection and storage, library storage, and combinatorial chemistry applications.
Información para pedidos: Plates with round-bottom wells are supplied 10 per sleeve, 12 sleeves per case. Plates with conical-bottom wells are supplied 20 per sleeve, 6 sleeves per case. For lids and sealing films, see 73520-478 series. Bar coding is available; ask your sales representative.