BioWhittaker® Reagents, Lonza
Proveedor: Lonza
BioWhittaker®
13-114E 17-839Z 17-838Z 13-607C 17-839Z 17-160E 17-711E 13-607C 17-829E 17-160E 17-161E 17-161E 13-114E 17-838Z 17-829E
CA12001-660 CA12001-634 CA12001-656 12002-030 CA12002-030 12001-718 12001-716 CA12001-704 CA12001-638 12001-638 CA12001-716 CA12001-718 12001-656 12001-634 12001-660
BioWhittaker® Reagents, Lonza
Suplementos y aditivos de cultivo celular
All BioWhittaker* reagents are manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility
All trypsin products are manufactured with irradiated trypsin, tested for porcine parvovirus, and screened for mycoplasma.