Full Height Sieves, Stainless Steel Wire and Frame, W.S. Tyler™
Proveedor: W.S. Tyler
5190 5214 5211 5205 5216 5200 5189 5193 5217 5208 5197 5210 5192 5215 5191 5209 5202 5203 5195 5196 5198 5212 5204 5199 5201 5213 5207 5194 5206
57324-080EA 3234.69 MXN
57324-080 57324-501 57324-545 57324-567 57324-523 57324-228 57324-206 57324-443 57324-421 57324-465 57324-487 57324-024 57324-002 57324-068 57324-046 57324-126 57324-148 57324-104 57324-400 57324-240 57324-320 57324-385 57324-363 57324-160 57324-181 57324-261 57324-283 57324-308 57324-272
Full Height Sieves, Stainless Steel Wire and Frame, W.S. Tyler™
Tamices Tamices de ensayo
Full height sieves are 5.08cm (2") high above stainless steel wire cloth
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number. Sieves one millimeter and coarser are identified by opening in millimeters and inches; sieves finer, by opening in microns and inches.
Conform to ASTM E-11 specifications.
Overall height: 7.4cm (2.9"). Diameter: 20.3cm (8").