VWR® Signature™ Pipette Tips for Rainin® LTS® Pipettors
Proveedor: VWR
VWR® Signature™
1011-960-306 1018-960-306 1155-965-306 1011-800-300 1146-225-300 1018-800-000 1018-290-320 1145-225-300 1015-250-300 1160-960-306 1145-965-306 1153-965-306 1019-960-306 1019-800-000 1153-800-300 1154-965-306 1066-800-300 1143-225-300 1068-260-300 1068-800-300 1144-965-306 1066-260-300
37001-608CS 8236.07 MXN
37001-608 83009-694 37001-604 83009-686 83009-698 37001-602 10015-686 83009-688 37001-600 37001-596 89495-382 82003-196 89495-384 89495-389 89495-422 89495-378 89495-400 82003-198 89495-376 89495-420 10015-704 89495-404
VWR® Signature™ Pipette Tips for Rainin® LTS® Pipettors
Puntas de pipetas
These clear polypropylene filter and non-filter resin tips fit Rainin® LTS® pipettors.
  • Ultrafine points
  • Autoclavable
  • Free of RNase, DNase, endotoxins, human DNA, and metals
The tips fit and eject with significantly less force than most conventional pipette tips. The natural barrier aerosol tips fit Rainin® LTS® pipettors and eject with significantly less force than conventional pipette tips. They feature ultrafine points and super clear resin for excellent viewing of sample.
Certificaciones: Lot certified.
Información para pedidos: Pak Rak® Express refills contain pipette tips to fill 8×12 racks, without the packaging waste of standard tip boxes. Pak Rak® Express refills require the Pak Rak® system loader, which dispenses tips quickly and easily from the refill pack into each rack, eliminating risk of contamination from hands or gloves.