Axygen® Sealing Films, Corning
Proveedor: Corning
Axygen®
UC-500 PCR-AS-200 BF-400-S PCR-SP-S PCR-AS-600 PCR-TS BF-400 PCR-TS-900 PCR-SP
47734-817CS 16712.16 MXN
47734-817 47743-943 89080-530 10011-121 47749-924 89080-528 10011-117 47749-926 10011-119
Axygen® Sealing Films, Corning
Juntas Sellos de microplaca
Sealing films are ideal for 96-well, 384-well, deep well, and other assay and PCR plates.
Aluminum Seal films maintain an excellent seal on microplates for a wide temperature range. Ideal for light-sensitive samples. Its uniformly applied adhesive provides optimal well sealing. When used in conjunction with Axygen compression mat, seals during thermocycling process. Color: reflective. Temperature range: –80 to 97°C (–112 to 207°F).
AxySeal films are polyester-based films with an acrylic adhesive ideal for ELISA and enzyme immunoassay applications. Color: clear. Temperature range: –40 to 65°C (–40 to 149°F).
UltraClear film is an exceptionally clear film specifically designed for meeting the optical requirements of Real-Time PCR. With a unique pressure-sensitive adhesive, this film has no tacky residue that can interfere with optical analysis. Color: clear. Temperature range: –40 to 65°C (–40 to 149°F).
Breathable films allow effective gas exchange for cellular/bacterial cultivation while preventing contamination. Color: clear. Temperature range: room temperature.