VWR® & VWR® Signature™ Bevel Point Pipette Tips for Eppendorf® Pipettors with Tip Ejector
Proveedor: VWR
Versapro
1167-800-000 1032-800-000 1032-965-306
53503-781CS 22373.63 MXN
53503-781 37001-182 53512-330
VWR® & VWR® Signature™ Bevel Point Pipette Tips for Eppendorf® Pipettors with Tip Ejector
Puntas de pipetas
These polypropylene tips are designed for fixed and variable volume pipettors that are equipped with a tip-ejector function.
  • Sterile tips are endotoxin-free
  • All tips are RNase- and DNase-free
  • Tips and hinged racks are fully autoclavable to 122 °C (252 °F)
Tips have a ribbed top and beveled end to ensure a precise fit and accurate volume delivery. Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA, and other viscous materials. Low-binding tips are also free of protease and human DNA.
Información para pedidos: Use Pak Rak® and Pak Rak Express refills in the Pak Rak system loader (see 30128-374 series) to reload pipette tip racks quickly and easily. Refills have greatly reduced packaging waste compared to standard tip boxes. Each Pak Rak refill contains one rack and tips to fill additional 8×12 racks. Pak Rak Express refills include only the tips, reducing waste even more.