VWR® & VWR® Signature™ Ultra-Micro Pipette Tips, Graduated
Proveedor: VWR
Versapro
1038-290-306 1038-965-336 1038-960-336 1161-800-000 1161-960-306 1038-800-000
53509-132CS 36941.91 MXN
53509-132 53509-130 37001-162 53503-810 37001-158 53509-070
VWR® & VWR® Signature™ Ultra-Micro Pipette Tips, Graduated
Puntas de pipetas
Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA and other viscous materials.
  • For use with most micropipettors
  • All tips are RNase- and DNase-free
  • Sterile tips are endotoxin-free
  • Tips and hinged boxes are Fully autoclavable to 122 °C (252 °F)
Tips have ultrafine, unbeveled points and are graduated at 2 and 10 µl. Low-binding tips are also protease- and human DNA-free.
Información para pedidos: Use Pak Rak® and Pak Rak Express refills in the Pak Rak system loader (see 30128-374 series) to reload pipette tip racks quickly and easily. Refills have greatly reduced packaging waste compared to standard tip boxes. Each Pak Rak refill contains one rack and tips to fill additional 8×12 racks. Pak Rak Express refills contain only the tips, reducing waste even more.