VWR® & VWR® Signature™ Genomic Pipette Tips
Proveedor: VWR
VWR®
1164-960-306 1164-965-306 1026-960-008-9
37001-186CS 15473.79 MXN
37001-186 37001-184 53503-612
VWR® & VWR® Signature™ Genomic Pipette Tips
Puntas de pipetas
These clear tips have an extra-large orifice to allow pipetting of fragile cells without shearing.
  • For use with most pipettors
  • Sterile tips are endotoxin-free
  • All tips are RNase- and DNase-free
  • Tips and hinged racks are autoclavable
Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA, and other viscous materials. Low-binding tips are also protease- and human DNA-free.