Whatman™ Antibiotic Assay Paper, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
2017-009 2017-013 2017-009 2017-006
89005-536PK 14700 MXN
89005-536 CA11013-048 89007-048 10035-734
Whatman™ Antibiotic Assay Paper, Whatman products (Cytiva)
Tests para microbiología Tests antibióticos
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method. The antibiogram allows rational and selective chemotherapy. The test disks are coated with chemotherapeutic agents, placed on the inoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
The antibiogram allows rational and selective chemotherapy. The test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.