Costar® Multiwell Microplates, Sterile, Corning®
Proveedor: Corning
Costar® Corning®
3738 3737 3737 3736
89093-498CS 2566.23 MXN
89093-498 89093-496 490014-862 89093-494
Costar® Multiwell Microplates, Sterile, Corning®
Microplacas Microplacas HTS
Clear, untreated polystyrene plates have a hydrophobic surface that is ideal when reduced cell attachment is desired
These flat bottom plates are supplied with nonreversible lids with condensation rings to reduce contamination. Plates feature individual alphanumeric codes to facilitate well identification, and a uniform footprint for easy stacking and storage. Sterilized by gamma irradiation. Certified nonpyrogenic. Bulk packed.