VWR® Gel Loading Pipet Tips
Proveedor: VWR, part of Avantor
1034-960-306 1034-800-000 1034-965-306
37001-152CS 22413.04 MXN
37001-152 37001-150 53509-015
VWR® Gel Loading Pipet Tips
Clear polypropylene tips include long, thin capillary sections for use in nucleic acid sequencing.
  • For use with most pipettors
  • Exclusive in hinged racks
The thin capillary fits between gel plates to enable accurate filling in between the teeth of the gel comb. Gel loading tips are 8.2 cm (3 1/4") in length with a 3 cm (13/16") capillary. Autoclavable. Certified RNase-, DNase-, and endotoxin-free.