VWR® Disposable Pipetting Reservoirs
Proveedor: VWR
VWR®
63303610 63303625 63303615 63303620 63303614 63303617 63303613 63303621 63303626 63303616 63303624 63303622 63303623 63303611 63303618 63303612 63303619
89094-656CS 3381.84 MXN
89094-656 89094-678 89094-668 89094-658 89094-674 89094-686 89094-664 89094-676 89094-666 89094-688 89094-670 89094-682 89094-660 89094-672 89094-662 89094-684 89094-680
VWR® Disposable Pipetting Reservoirs
Depósitos de reactivos
VWR® Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width.
  • Available in distinct 25, 50, or 100 mL formats
  • Available sterile or nonsterile
  • Lot-certified, nonpyrogenic, DNase- and RNase-free
Pipetting reservoirs feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.
The 25 mL reservoirs are also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 mL capacity. The 50 mL reservoirs are available in clear FDA grade PVC or virgin modified polystyrene. All other sizes are manufactured from virgin modified polystyrene. Available nonsterile or electron beam sterilized.