VWR® Clinical 200 Large Capacity Centrifuges
Proveedor: VWR
VWR®
C0200-AC-VWR
82017-654EA 127326.25 MXN
82017-654
VWR® Clinical 200 Large Capacity Centrifuges
Centrífugas Centrífugas de sobremesa
Designed for ease of use, this large-capacity centrifuge is ideal for clinical lab and research environments.
  • For clinical lab and research environments
  • Microprocessor control
  • Safety lock lid
  • Dynamic braking with non-disruptive soft stop
  • VWR® Two-Year Limited Parts and Labor Warranty
Versatile unit accepts tubes from 1.5 mL to 50 mL, including all common blood draw tubes
A combination of digital displays and easy-set knobs allow for quick and precise setting of operating parameters. Precise operational control is maintained by the microprocessor. An imbalance sensor is incorporated into control the loop to stop operation of centrifuge in case rotor is loaded improperly. At end of run, dynamic braking brings rotor to quick and turbulence-free stop.
Four rotor options are available. Optional adapters accommodate various tube sizes including standard round-bottom blood tubes. Safety lock lid prevents opening during use. Centrifuge may be used in a cold room. Hematocrit tube reader measures hematocrit values with tubes still in rotor; device is placed on top of rotor and corresponding value is read from graph.
Hematocrit tube rotor is also available.
Certificaciones: CE marked.
Información para pedidos: Centrifuges 82017-654 and -656 include a 28° fixed-angle rotor that holds twelve 15 mL tubes and adapters for 5 mL, 7 mL, and 10–12 mL tubes.