VWR® Clinical 50 Centrifuge
Proveedor: VWR
VWR®
C0050-A-VWR
82013-800EA 24160.54 MXN
82013-800
VWR® Clinical 50 Centrifuge
Centrífugas Centrífugas de sobremesa
Centrifuge features a window in the lid for viewing the rotor.
  • For physicians' offices and small laboratories
  • Clear lid with safety interlock
  • VWR® Two-Year Limited Parts and Labor Warranty
A safety interlock cuts power to the centrifuge when the lid is opened
Supplied complete with a 6x15 mL fixed-angle rotor, this basic unit features variable speed to 4000rpm/1900g, suitable for a variety of applications in the clinical setting or research lab. The rotor accepts 15 mL (16x130mm) and 10 mL (16x100mm) tubes directly. Optional adapters accommodate smaller tubes, including common blood draw tubes.
The rugged ABS housing adds durability to the centrifuge and is easy to clean.