VWR® Modular Heating Blocks for Vials
Proveedor: VWR
VWR®
80863213 80863210 80863209 80863212 80863215 80863214 80863208 80863207 80863211
12621-134EA 3672.9 MXN
12621-128 12621-126 12621-138 12621-124 12621-136 12621-134 12621-132 12621-130 12985-066
VWR® Modular Heating Blocks for Vials
Termobloques Bloques
Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.