VWR® Maxi 20 Electrophoresis System
Proveedor: VWR
VWR®
E1020-20-VWR
89032-298EA 20899.87 MXN
89032-298
VWR® Maxi 20 Electrophoresis System
Sistemas de electroforesis Sistemas horizontales de electroforesis en gel
Ideal for processing large numbers of samples, including PCR products from 96-well plates, RFLP analysis, or plasmid screenings.
  • High-throughput screening
  • Will withstand everyday use
Gel box and tray are molded with thick acrylic for leakproof performance
Durable rubber casting gates permit easy, tape-free casting, while slots in the sides of the trays allow for easy comb placement. Two-piece comb design provides for vertical adjustment, giving control over well depth. Platinum electrode cassettes remove easily for cleaning or replacement.
The "EasyLift" gel box lid is easily removed using the side tabs and pressure pads. Domed lid prevents condensation from dripping onto gel and is surrounded by a drip ring to help recover the condensate and maintain buffer concentration.
Tray is UV transparent for easy visualization of samples. System accepts 20 x 20 cm trays. Corrosion-resistant electrodes are held in protective cassettes for safe cleaning and easy replacement. System provides for high-throughput screening of samples, high resolution, and is built to withstand rigors of everyday use.
Información para pedidos: Supplied with one 20 x 20 cm gel tray; two 20-tooth, 1.5 mm thick combs; two 40-tooth, 1.5 mm thick combs; casting dams; and well visualization strips.