VWR® Midi Plus Horizontal Electrophoresis Systems
Proveedor: VWR
VWR®
E1015-15-VWR E1015-10-VWR
89032-294EA 15302.95 MXN
89032-296 89032-294
VWR® Midi Plus Horizontal Electrophoresis Systems
Sistemas de electroforesis Sistemas horizontales de electroforesis en gel
These two versatile systems are ideal for resolving close molecular weight samples and restriction fragments.
  • Versatile systems
  • Multichannel combs
  • Quick and easy loading
Multichannel combs permit quick and easy loading of large numbers of samples.
Gel boxes and trays are molded with thick acrylic for leakproof performance. Trays are UV transparent for easy visualization of samples. Systems accept both 15 x 10 cm and 15 x 15 cm gel trays. Corrosion-resistant platinum electrodes are held in protective cassettes for safe cleaning and easy replacement.
Durable rubber casting gates permit easy, tape-free casting, while slots in the sides of the trays allow for easy comb placement. Two-piece comb design provides for vertical adjustment, giving control over well depth.
The EasyLift" gel box lid is easily removed using the side tabs and pressure pads. Domed lid prevents condensation from dripping onto gel and is surrounded by a drip ring to help recover the condensate and maintain buffer concentration.
Información para pedidos: Midi Plus 10 gel system is supplied with one 15 x 10 cm gel tray; one 16-tooth, 1.5 mm thick comb; one 20-tooth, 1.5 mm thick comb; casting dams; and well visualization strips. Midi Plus 15 gel system is supplied with one 15 x 15 cm gel tray; one 16-tooth, 1.5 mm thick comb; one 20-tooth, 1.5 mm thick comb; one 28-tooth, 1.5 mm thick comb; casting dams; and well visualization strips.