VWR® Clinical 100 Laboratory Centrifuge
Proveedor: VWR
VWR®
C0060-VWR
97011-432EA 32386.39 MXN
97011-432
VWR® Clinical 100 Laboratory Centrifuge
Centrífugas Centrífugas de sobremesa
Centrifuge is designed for a wide variety of applications in clinical and research laboratories.
  • Ideal for PRP (Platelet Rich Plasma) and PPP (Platelet Poor Plasma) applications
  • Digital speed and time control
  • Lid with safety interlock
  • VWR® Two-Year Limited Parts and Labor Warranty
It is supplied complete with rotor and accepts 15 ml conical and 10 ml (16×100 mm) tubes directly, and smaller tubes with the use of an optional adapter.
The control panel is equipped with easy-to-turn knobs and large digital displays for time and speed. During operation the centrifuge lid is locked, and the chamber cannot be accessed until the run is complete. The unit's compact footprint allows it to fit easily within crowded workspaces, and it can be operated on benchtops or in cold rooms. The rugged, molded housing provides a long service life and is easy to clean.
With variable speed control in increments of 100 rpm, the unit is ideal for producing platelet rich plasma (PRP) for agglutination studies, urine sedimentation, chemistry analyte sample preparation, and pelleting of cultures. The high speed capabilities allow for quick production of platelet poor plasma (PPP) for coagulation studies.
Información para pedidos: The optional combination tube adapter (82013-820) accommodates 5 ml (12×75 mm) and 7 ml (13×100 mm) tubes.