Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2117-1000 2117-0060 2117-0125 2117-0250 2117-0500
16129-378CS 8360.91 MXN
16129-390 16129-414 16129-354 16129-356 16129-378
Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Tarros Tarros para uso general
Transparent, autoclavable, with straight sides
Top is recessed to allow stacking. Jars of 125mL and 250mL sizes have standard mason jar threads. Ideal for museum jars — formaldehyde loss only about 2% per year. Remove closure before autoclaving, or set closure on top of jar at an angle without engaging the threads.
Suitable for food and beverage use. Meets requirements of CFR21, Part 177.1520 section of Food Additives Amendment of Federal Food and Drug Act.
Polypropylene white screw closures are linerless, noncontaminating, and autoclavable.