Nalgene® Polypropylene Straight-Sided Jars, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2118-0004 2118-0032 2118-0008 2118-9050 2118-0016 2118-0002 2118-0001
36319-627CS 6412.47 MXN
36319-627 36319-501 36319-547 36319-503 36319-707 36319-580 36319-660
Nalgene® Polypropylene Straight-Sided Jars, Thermo Scientific
Tarros Tarros para uso general
For storing and collecting lab supplies and specimens
Suitable for food and beverage use. Meets requirements of CFR21, Part 177.1520 section of Food Additives Amendment of Federal Food and Drug Act.
Translucent with excellent chemical resistance. With natural polypropylene linerless closures. Jars of 125mL and 250mL sizes fit blenders with mason jar threads. Remove closure before autoclaving, or set on top of jar at an angle without engaging threads. Repeatedly autoclavable.