Nalgene® Centrifuge Bottles with Caps, Polycarbonate, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
3122-0500 3140-1006 3140-0500 3122-1000 3122-1010 3140-0250 3122-0250
21020-108CS 22823.26 MXN
21020-094 89050-118 21020-108 21020-130 21020-163 21010-432 21010-421
Nalgene® Centrifuge Bottles with Caps, Polycarbonate, Thermo Scientific
Frascos Frascos de centrífuga
Wide mouth bottles are transparent, with excellent mechanical strength.
Screw cap for bottle 21020-108 can be replaced with sealing cap assembly 21009-901 for leakproof performance at high speeds or when hazardous materials must be contained.
Note: Before autoclaving, place cap on container without engaging threads. For proper performance, fill bottles to at least 80% of the total capacity.
All caps are polypropylene except the cap for bottle 89050-118, which is modified polyphenylene-ether. Sealing caps feature a retained silicone gasket. All bottles and caps are autoclavable.