Gas, Air, or Vacuum Fittings, Deck Mounted, WaterSaver Faucet
Proveedor: WaterSaver Faucet
L4200FH132AW L4200131WSA L4200FH134WS L4200FH132SW
97004-550EA 4556.26 MXN
97004-548 97004-546 97004-552 97004-550
Gas, Air, or Vacuum Fittings, Deck Mounted, WaterSaver Faucet
Grifos y fregaderos Grifos
These deck-mounted faucets are made of chrome-plated brass.
  • Maximum Working Pressure: 75 psi
  • Index Buttons for Gas, Air, and Vacuum
  • Single, Double, or Quadruple Service Available
  • Chrome-Plated Brass Construction
The fittings consist of an outlet turret and ball valve.
The faucet includes index buttons for gas, air, and vacuum, and single, double, or quadruple service is available.
Información para pedidos: This item includes 9.5 mm, (3/8") IPS tank nipples, locknuts, and washers.