MLS Series Top-Loading Portable Autoclaves, Panasonic Healthcare
Proveedor: Panasonic
MLS3751LPA
97002-402EA 426075 MXN
97002-402
MLS Series Top-Loading Portable Autoclaves, Panasonic Healthcare
Esterilizadores Autoclaves (Esterilizadores de vapor)
The importance of sterilization is essential to good laboratory practice, particularly in biotechnology, pharmaceutical, and clinical laboratories.
  • Twelve preset programs available
Of all sterilization methods, high-pressure steam (autoclave) is the most widely used due to its efficacy, speed, and reliability
They allow maximum flexibility for the sterilization of laboratory equipment and media. A compact design allows for flexible placement in any facility while the large chamber offers capacity for up to eight 1 L flasks. 12 preset programs are available as well as independent programming that allows users to set their own programs. The PHCbi top-loading portable autoclave maximizes the use of available floor space while still providing ample capacity for storage within the 37 cm (14.6") chamber.
The PHCbi MLS Series top-loading autoclaves provide a safe and reliable high-pressure steam sterilizing environment within an easy-to-use, self-contained unit. These energy-saving autoclaves are ideal for a wide range of applications.
Certificaciones: QPS listed. RoHS compliant.
Información accesorios: The object temperature sensor can be installed to measure the temperature of items in the autoclave chamber and to set the sterilization timer. This sensor is suitable for materials, such as resins, that heat up more slowly than the chamber. The object temperature sensor must be purchased separately.
The thermal printer can be installed for printing date- and time-stamped process run results: temperature, pressure, and time. The printer must be purchased separately.
Información para pedidos: All units are supplied with a one-year manufacturer's parts and labor warranty, drain hose, exhaust tank, exhaust hose, tank mounting bracket, and NEMA locking plug. Model MLS3751L is supplied with two stainless steel wire mesh baskets. Model MLS3781L is supplied with three stainless steel wire mesh baskets. Accessories are supplied with a one-year manufacturer's warranty.