SP Bel-Art PiRack® Pipettor Holder System, Bel-Art Products, a part of SP
Proveedor: Bel-Art
Pi Rack™
F18954-0002 F18954-0001 F18954-0500 F18954-0000
47727-254EA 1240.52 MXN
47727-254 47727-262 47727-252 47750-510
SP Bel-Art PiRack® Pipettor Holder System, Bel-Art Products, a part of SP
Soportes de pipetas
This polypropylene system is strong enough to support the weight of even the heaviest pipettor.
  • Polypropylene construction
  • Strong and durable
  • Utility tray included for holding small pipetting supplies
  • Chemical-resistant
  • Autoclavable
Hold a single pipettor with one clamp, or add a triple clamp or extenders to increase the number of holders at a work location. The utility tray, which mounts between two clamps, is ideal for storing pipetting supplies or other small items.
Attach to any shelf or benchtop edge up to 2.54 cm (1") thick using the large locking thumbscrew clamp. Supports many standard brands of pipettors. All components are chemical-resistant and autoclavable at 121°C (250°F).