VWR® Pipettor Holders
Proveedor: VWR
VWR®
HS20162AV
82007-226CS 36499.28 MXN
82007-226
VWR® Pipettor Holders
Soportes de pipetas
Autoclavable polypropylene holders keep pipettors at your fingertips.
  • Polypropylene construction
  • Autoclavable
  • Adjustable ABS plastic screws
Adjustable ABS plastic screws allow secure attachment to table tops and benches up to 3 cm (13/16") thick. Accommodate most brands of pipettors.