KV3000 Digital Constant Temperature Kinematic Viscosity Baths, Koehler
Proveedor: Koehler Instrument
K23706 K23796 K23700
82017-698EA 320973.41 MXN
37000-166 82017-698 37000-170
KV3000 Digital Constant Temperature Kinematic Viscosity Baths, Koehler
Baños Baños de viscosímetro
Constant temperature bath series with advanced temperature control circuitry and integrated timing features for convenient, accurate glass capillary viscometry determinations
Microprocessor PID circuitry assures precise, reliable temperature control within ASTM-specified tolerances throughout the operating range of the bath. Simple push-button controls and dual digital displays permit easy setting and monitoring of bath temperature. Two-place calibration offset capability is provided. Baths accommodate seven glass capillary viscometers of various types. Viewing the viscometers is made easy by glare-free fluorescent illumination inside the bath and a baffle that provides a background for easy viewing. Temperature control uniformity is assured by means of motorized stirrer that provides complete circulation without turbulence. Connection of the built-in cooling coil to tap water or a recirculating water chiller facilitates temperature control at ambient or below ambient temperatures.
Certificaciones: Conforms to the specifications of: ASTM D445, D2170, D6074, D6158; IP 71, 319; ISO 3104; DIN 51500; FTM 791-305; NF T 60-100.
Información para pedidos: For information about communications software, ramp-to-set, and other enhanced features, contact your Avantor sales representative.