VWR® Disposable Bacteriological Pipettes, Glass, Sterile, Plugged
Proveedor: Avantor
VWR®
72108-10110 72108-5110
53281-816CS 8796.94 MXN
53281-816 53281-818
VWR® Disposable Bacteriological Pipettes, Glass, Sterile, Plugged
Pipetas Pipetas bacteriológicas
Wide tips are excellent for transferring viscous or suspended liquids.
  • Provided with negative graduations
  • Excellent for transferring viscous or suspended liquids
  • Glass fabrication
  • Sterile
  • Plugged
Calibrated to deliver to the tip. Two rings near the top denote that the small amount remaining in the tip after free delivery must be blown out and added to the main volume delivered. Provided with negative graduations to increase the versatility of the pipette.
Información para pedidos: Supplied in multi-pack bags to protect against contamination.