Shielded Compact Flowmeters, Gilmont®, Cole Parmer
Proveedor: Cole Parmer
03232-22 03232-20 323223
29895-242 29895-264 29895-209
Shielded Compact Flowmeters, Gilmont®, Cole Parmer
Flowmeters can be applied to systems under pressure or under conditions where exposed glass may create a hazard.
Flowmeters feature protective all-plastic shields that confine fluid contact to PTFE, Viton®, and glass only. Fluid does not contact polypropylene bushing or protective plastic tube. Shields employ plastic materials for high chemical resistance. Outer protection tube made of polycarbonate with excellent optical properties. Bushings made of corrosion-resistant polypropylene with PTFE inserts. Standard pipe thread on ends and PTFE insert will also take flexible hose. O-ring seal ensures gas-tight connection. With direct reading scales of 100mm in length, they are accurate to ±5% or ±2mm of scale length (whichever is greater). Each meter supplied with direct reading air and water scales.