Anaerobic Systems, BD Diagnostics
Proveedor: BD Diagnostics
260413 260467 260619 260628 260629 260609 260618 260467 260463 260627 260607 260413 270124 260638 260618 260610 260411 260610 260626 260463 260627 260608 260628 260609 260637 260414 260414 260607 260626 260608 260411 260614 260629 260614
90003-606EA 21812.35 MXN
90003-608 90003-604 90003-606 90003-588 90003-822 90003-602 CA90003-594 90003-584 90003-586 90003-564 90003-592 90003-594 CA90003-580 90003-590 CA90003-606 CA90003-608 90003-614 CA90003-602 CA90003-604 CA90003-564 CA90003-586 90003-612 CABB260414 CABB260413 CA90003-588 CA90003-582 90003-596 CA90003-584 CA90003-590 90003-580 BB4360413 90003-582 BB4360414 CA90003-592
Anaerobic Systems, BD Diagnostics
Sistemas anaeróbicos
Designed for the generation of anearobic, microaerophilic (reduced oxygen), or CO2-enriched environments.
The BD GasPak™ 100 and 150 self-contained anaerobic systems consist of one polycarbonate jar (vented or non-vented), lid with O-ring gasket, catalyst reaction chamber, BBL room temperature catalyst charges, one rack, and one tube holder. The BD GasPak™ 100 system features two catalyst charges; the BD GasPak™ 150 system features ten charges. These systems use hydrogen and carbon dioxide generator envelopes and do not require a vacuum pump, gas tank, manometer, or pressure reducing valve. The BD GasPak™ 100 system jar holds up to twelve 100mm (315/16") dishes. The BD GasPak™ 150 system jar holds up to twelve 150mm (515/16") petri dishes or thirty-six 100mm (315/16") dishes. The BD GasPak™ 150 vent kit (90003-588) consists of a threaded vent, rubber tube, and a tubing clamp for converting a non-vented system into a vented system.
Información para pedidos: For more components and accessories, contact your Avantor sales representative.