Fendall® Saline Eyewash Wall Stations, Honeywell Safety
Proveedor: Honeywell Safety
Fendall®
32-000462-0000-H5 32-000460-0000-H5 32-000461-0000
56611-130CS 6691.04 MXN
56611-130 56611-126 56611-128
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
Suministros de primeros auxilios Estaciones de lavado de ojos
Available in 470 ml (16 oz.) and 950 ml (32 oz.) single and 950 ml (32 oz.) double wall stations.
  • Two-sided tape and screws are provided for mounting
  • Classified as personal/secondary eyewash device
Bottles have extended flow openings and twist-off tops for easy access in an emergency. Each bottle is expiry dated and lot numbered for end-user protection.