Deck-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Proveedor: WaterSaver Faucet
L412-12 L414 L412-8 L411 L411-8
VFL412-8EA 8312.49 MXN
VFT-0340-00 VFL412 VFL412-8 VFL412-12 59382-042
Deck-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Grifos y fregaderos Grifos
Deck-mounted mixing faucets are made of chrome-plated forged brass and provided with a serrated hose end, four-arm handles, and 1cm (3/8") diameter copper tubes on inlets.
Faucets are available with a 15.2 cm (6"), 20.3 cm (8"), or 30.5 cm (12") spread gooseneck. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certificaciones: CSA certified. Comply with ANSI/ASME A112.18.1M.