Cleanroom Talk-Through, Terra Universal
Proveedor: Terra Universal
5050-50
23000-530EA 9984.58 MXN
23000-530
Cleanroom Talk-Through, Terra Universal
Habitaciones prefabricadas
Provides clear communication and visibility
Ideal for cleanrooms, laboratories, and hospital isolation, the cleanroom talk-through mounts on a 19.7cm (73/4") diameter cutout. The overlapping Mylar® S-shaped diaphragms allow clear communication and visibility. Unit weight: 0.91kg (2lbs.).
Información para pedidos: Custom installation panels are available for other cut-out requirements; contact your Avantor Furniture Specialist for complete information.